Imprimer
Bovine elastins (from neck ligament), MP Biomedicals
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0210163610 0210163601 0210163605 0210163625
ICNA0210163610EA 195 EUR
ICNA0210163610 ICNA0210163601 ICNA0210163605 ICNA0210163625
Bovine elastins (from neck ligament), MP Biomedicals
Protéines et Peptides
Elastin is highly stable and hemocompatible protein now finding use in construction of biomaterials as a result of several strategies for its solubilisation. It is a structural protein which has been used in studies of Chronic obstructive pulmonary disease (COPD). It may be used to investigate why there is a lack of repair for these proteins in patients with COPD.