Imprimer
NMR tubes, high quality grade, KIMBLE®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SP897325-0000 SP897320-0000
SCERSP897325-0000EA 77.5 EUR
SCERSP897325-0000 SCERSP897320-0000
NMR tubes, high quality grade, KIMBLE®
Tubes Tubes RMN
Borosilicate glass, 51 expansion, Type I and ASTM E438, Type I, Class B.
  • With attached polyethylene caps
  • These HQ NMR tubes are highest quality grade and are 100% gauged for wall thickness, concentricity and camber specifications.
External diameter: 10 mm
Internal diameter: 9,07 mm
Wall thickness: 0,46 mm