(Methoxycarbonylsulphamoyl)triethylammonium hydroxide inner salt 96%
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Attention
|
Synonyme(s):
Methyl N-(Triethylammoniosulphonyl)carbamate, Burgess Reagent
Purity: 96%
CAS No.: 29684-56-8
Molecular Formula: C8H18N2O4S
Formula Weight: 238.30
Storage Temperature: -30°C to -10°C
Melting Point: 72-82°
Beilstein Reference: 1432131
MDL No.: MFCD00077815
Formule:
C₈H₁₈N₂O₄S Point de fusion: 72…82 °C |
Numéro MDL:
MFCD00077815 Numéro CAS: 29684-56-8 |