Imprimer

Carbimazole 97%

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Attention

Synonyme(s): 1-Ethoxycarbonyl-3-methyl-2-thioxo-4-imidazoline, Ethyl 3-methyl-2-thioxo-2,3-dihydro-1H-imidazole-1-carboxylate

L08218.14 L08218.22
L08218.14EA 221 EUR
L08218.14 L08218.22
Carbimazole 97%
Carbimazole

Purity: 97% CAS No.: 22232-54-8 Molecular Formula: C7H10N2O2S Formula Weight: 186.24 pH: 5.0-7.0 Melting Point: 122-126° Beilstein Reference: 144339 MDL No.: MFCD00027421

Formule: C₇H₁₀N₂O₂S
Point de fusion: 122…126 °C
Température de stockage: Température ambiante
Numéro MDL: MFCD00027421
Numéro CAS: 22232-54-8
EINECS: 244-854-4
Indice Merck: 14,1798

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR