Imprimer

Reserpine 99%

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Attention

L03506.03 L03506.06
L03506.03EA 106 EUR
L03506.03 L03506.06
Reserpine 99%
Reserpine

Purity: 99% CAS No.: 50-55-5 Molecular Formula: C33H40N2O9 Formula Weight: 608.69 Melting Point: ca 265° Beilstein Reference: 102014 Optical Rotation: -122° (c=1 in chloroform) MDL No.: MFCD00005091

Formule: C₃₃H₄₀N₂O₉
Point de fusion: ∼ 265 °C
Température de stockage: Température ambiante
Numéro MDL: MFCD00005091
Numéro CAS: 50-55-5
EINECS: 200-047-9
Indice Merck: 14,8145

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR