Imprimer

Cefuroxime sodium salt 95%

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Danger

J66999.03 J66999.06
J66999.03EA 183 EUR
J66999.03 J66999.06
Cefuroxime sodium salt 95%
Cefuroxime sodium salt

Purity: 95% CAS No.: 56238-63-2 Molecular Formula: C16H15N4NaO8S Formula Weight: 446.37 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to pale yellow Melting Point: 240-245° Solubility: Freely soluble in water Optical Rotation: +60° (c=1 in water) MDL No.: MFCD09878727

Formule: C₁₆H₁₅N₄NaO₈S
Point de fusion: 240…245 °C
Numéro MDL: MFCD09878727
Numéro CAS: 56238-63-2
EINECS: 260-073-1
Indice Merck: 14,1954

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR