Imprimer

Bialaphos sodium ≥85%

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Danger

J66802.MC J66802.ME
J66802.MCEA 301 EUR
J66802.MC J66802.ME
Bialaphos sodium ≥85%
Bialaphos sodium

Has successfully been used in major cereal species transformation.


Applications: Bialaphos Sodium Salt is a tripeptide antibiotic that is employed in the transformation experiments of many species of plants that make use of the bar gene for selection. It is more effective than glufosinate in corn. In wheat, bialaphos was the most reliable selection criteria used to identify bar transformed plants.


Applications: Has successfully been used in major cereal species transformation


Attention: Notes: Research Use only: Not intended for animal or human diagnostic or therapeutic use

Formule: C₁₁H₂₁N₃NaO₆P
Point d'ébullition: 793 °C
Point de fusion: 160 °C
Point éclair: >110 °C (230 °F)
Numéro MDL: MFCD01674432
Numéro CAS: 71048-99-2
ADR: 6.1,III

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR