Colimycin M
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s):
Polymyxin E sodium methanesulphonate, Colistin sodium methanesulphonate
CAS No.: 8068-28-8
Molecular Formula: C58H115N16Na5O28S5
Formula Weight: 1759.89
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to pale yellow
Solubility: Freely soluble in water
MDL No.: MFCD00130824
Applications: Has a bactericidal effect by targeting the cell membrane and modifying its permeability
Formule:
C₅₈H₁₀₅N₁₆Na₅O₂₈S₅ Poids moléculaire: 1749.811 g/mol |
Numéro MDL:
MFCD00130824 Numéro CAS: 8068-28-8 EINECS: 232-516-9 |