Imprimer

Ribostamycin sulphate 630 µg/mg

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Danger

J66155.MD J66155.03
J66155.MDEA 272 EUR
J66155.MD J66155.03
Ribostamycin sulphate 630 µg/mg
Ribostamycin sulphate

A broad spectrum antibiotic targeting both Gram-positive and Gram-negative bacteria.


  • White to pale yellow powder


Soluble in water.


Attention: Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Formule: C₁₇H₃₄N₄O₁₀·H₂SO₄
Point de fusion: 175…180 °C
Point éclair: >110 °C (230 °F)
Température de stockage: Réfrigérateur
Numéro MDL: MFCD00057496
Numéro CAS: 53797-35-6
EINECS: 258-783-1
Indice Merck: 14,8206

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR