Nα-(p-Toluenesulphonyl)-DL-lysine chloromethyl ketone hydrochloride 98%
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Attention
|
Purity: 98%
CAS No.: 4238-41-9
Molecular Formula: C14H21ClN2O3S.HCl
Formula Weight: 369.31
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Off-white
Solubility: Soluble in water
MDL No.: MFCD00012513
Applications: Inhibitor of papain and trypsin
Formule:
C₁₄H₂₂Cl₂N₂O₃S |
Numéro MDL:
MFCD00012513 Numéro CAS: 4238-41-9 EINECS: 224-199-0 |