Fexofenadine hydrochloride
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s):
2-(4-(1-Hydroxy-4-(4-(hydroxydiphenylmethyl)piperidin-1-yl)butyl)phenyl)-2-methylpropanoic acid hydrochloride, Fexofenadine HCl
CAS No.: 153439-40-8
Molecular Formula: C32H39NO4.HCl
Formula Weight: 538.12
Storage Temperature: -30°C to -10°C
Physical Form: Crystalline powder
Appearance: White
Melting Point: 148-150°
MDL No.: MFCD00865710
Applications: H-1 receptor antagonist
Formule:
C₃₂H₃₉NO₄ Point de fusion: 148…150 °C |
Numéro MDL:
MFCD00865710 Numéro CAS: 153439-40-8 Indice Merck: 14,4068 |