Adenosine-3',5'-cyclic monophosphate sodium salt 99%
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s):
3′,5′-Cyclic AMP sodium salt, 3',5'-Cyclic AMP Sodium Salt, Adenosine 3',5'-Cyclic Monophosphate Sodium Salt Hydrate, 3',5'-cAMP Na, (4aR,6R,7R,7aS)-6-(6-Amino-9H-purin-9-yl)-2,7-dihydroxytetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinine 2-oxide, sodium salt, cAMP-Na, 3',5'-Cyclic AMP Sodium Salt Hydrate, 3',5'-cAMP Na Hydrate
Purity: 99%
CAS No.: 37839-81-9
Molecular Formula: C10H11N5NaO6P
Formula Weight: 351.19
Storage Temperature: -30°C to -10°C
Physical Form: Crystalline powder
Appearance: White
Beilstein Reference: 54612
MDL No.: MFCD00069736
Formule:
C₁₀H₁₁N₅NaO₆P Poids moléculaire: 351.19 g/mol |
Numéro MDL:
MFCD00069736 Numéro CAS: 37839-81-9 |