Agarose gel loading dye 6X for electrophoresis, alkaline
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Agarose gel loading dye (6X), alkaline is used in DNA gel analysis in alkaline condition.
- 300 mm sodium hydroxide
- 6 mm EDTA
- 0,25% bromocresol green
- 0,25% xylene cyanol FF
- 18% Ficoll®
Used in agarose gel electrophoresis.