Imprimer

Nimodipine ≥98%

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Attention

Synonyme(s): 3-Isopropyl 5-(2-methoxyethyl) 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate, Isopropyl 2-Methoxyethyl 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate

J61287.ME J61287.03
J61287.MEEA 123 EUR
J61287.ME J61287.03
Nimodipine ≥98%
Nimodipine

Purity: 98+% CAS No.: 66085-59-4 Molecular Formula: C21H26N2O7 Formula Weight: 418.44 Physical Form: Powder Appearance: Pale yellow MDL No.: MFCD00153848


Applications: Potent L-type calcium channel antagonist

Formule: C₂₁H₂₆N₂O₇
Température de stockage: Température ambiante
Numéro MDL: MFCD00153848
Numéro CAS: 66085-59-4
EINECS: 266-127-0
Indice Merck: 14,6551

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR