Imprimer

Ethyl 2-aminonicotinate 98%

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Attention

Synonyme(s): Ethyl 2-aminonicotinate

H64957.06 H64957.14 H64957.22
H64957.06EA 55.3 EUR
H64957.06 H64957.14 H64957.22
Ethyl 2-aminonicotinate 98%
Ethyl-2-aminonicotinate

Purity: 98% CAS No.: 13362-26-0 Molecular Formula: C8H10N2O2 Formula Weight: 166.18 Physical Form: Solid Melting Point: 90-96° lit. Beilstein Reference: 124680 MDL No.: MFCD01569452

Formule: C₈H₁₀N₂O₂
Point de fusion: 90…96 °C (lit.)                      
Température de stockage: Température ambiante
Numéro MDL: MFCD01569452
Numéro CAS: 13362-26-0

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR