Imprimer

8-Bromo-1-octène 97%

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Attention

H64830.03 H64830.06
H64830.03EA 59.1 EUR
H64830.03 H64830.06
8-Bromo-1-octène 97%
8-Bromo-1-octène

Purity: 97% CAS No.: 2695-48-9 Molecular Formula: C8H15Br Formula Weight: 191.11 Flash Point: 78°(172°F) Physical Form: Liquid Density: 1.139 Refractive Index: 1.4670 Appearance: Pale yelllow Boiling Point: 92°/24mm Beilstein Reference: 2234481 MDL No.: MFCD00000275

Formule: C₈H₁₅Br
Point d'ébullition: 92 °C (24 mmHg)
Densité: 1,139
Point éclair: 78 °C (172 °F)
Température de stockage: Température ambiante
Numéro MDL: MFCD00000275
Numéro CAS: 2695-48-9
EINECS: 220-268-4

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR