Imprimer

Acide 5-bromo-2-iodobenzoïque 97%

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Attention

H64643.06 H64643.14
H64643.06EA 59.7 EUR
H64643.06 H64643.14
Acide 5-bromo-2-iodobenzoïque 97%
Acide 5-bromo-2-iodobenzoïque

Purity: 97% CAS No.: 21740-00-1 Molecular Formula: C7H4BrIO2 Formula Weight: 326.92 Physical Form: Solid Sensitivity: Light Sensitive Melting Point: 158-163° Beilstein Reference: 3247380 MDL No.: MFCD00079717

Formule: C₇H₄BrIO₂
Point de fusion: 158…163 °C
Température de stockage: Température ambiante
Numéro MDL: MFCD00079717
Numéro CAS: 21740-00-1

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR