Benzyl-3-oxopiperidine-1-carboxylate 97+%
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Attention
|
Purity: 97+%
CAS No.: 61995-20-8
Molecular Formula: C13H15NO3
Formula Weight: 233.27
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Beilstein Reference: 1464624
MDL No.: MFCD03001723
Formule:
C₁₃H₁₅NO₃ |
Numéro MDL:
MFCD03001723 Numéro CAS: 61995-20-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...