3,6-Dihydro-2H-pyran-4-boronic acid pinacol ester 98%
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Attention
|
Synonyme(s):
3,6-Dihydro-2H-pyran-4-boronic acid Pinacol ester, 2-(3,6-Dihydro-2H-pyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,6-Dihydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-pyran, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyran
Purity: 98%
CAS No.: 287944-16-5
Molecular Formula: C11H19BO3
Formula Weight: 210.08
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Sensitivity: Moisture Sensitive
Appearance: White to off-white
Melting Point: 63-67°
Beilstein Reference: 8548328
MDL No.: MFCD11052631
Formule:
C₁₁H₁₉BO₃ Poids moléculaire: 210.08 g/mol Point de fusion: 63…67 °C |
Numéro MDL:
MFCD11052631 Numéro CAS: 287944-16-5 |