2-(3-Trifluoromethylphenyl)ethylamine 98%
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Danger
|
Synonyme(s):
3-(Trifluoromethyl)phenethylamine
Purity: 98%
CAS No.: 52516-30-0
Molecular Formula: C9H10F3N
Formula Weight: 189.18
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Density: 1.18
Refractive Index: 1.4645
Sensitivity: Light Sensitive
Appearance: Colourless to pale yellow
Boiling Point: 113°/22mm
Beilstein Reference: 4398117
UN No.: UN2735
MDL No.: MFCD00040756
Formule:
C₉H₁₀F₃N Point d'ébullition: 113 °C (22 mmHg) Densité: 1,18 |
Numéro MDL:
MFCD00040756 Numéro CAS: 52516-30-0 EINECS: 257-988-3 ADR: 8,III |