Imprimer

2,2-dimethylbut-3-enoic acid 95%

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Danger

Synonyme(s): 2,2-Dimethylbut-3-enoic acid

H64067.03 H64067.06
H64067.03EA 198 EUR
H64067.03 H64067.06
2,2-dimethylbut-3-enoic acid 95%
2,2-dimethylbut-3-enoic acid

Purity: 95% CAS No.: 10276-09-2 Molecular Formula: C6H10O2 Formula Weight: 114.14 Storage Temperature: 2°C to 8°C Physical Form: Liquid Density: 0.957 Refractive Index: 1.4295 Appearance: Colourless Melting Point: -6° Boiling Point: 185° Beilstein Reference: 1700758 UN No.: UN3265 MDL No.: MFCD00196967

Formule: C₆H₁₀O₂
Poids moléculaire: 114.144 g/mol
Point d'ébullition: 185 °C
Point de fusion: –6 °C
Densité: 0.957
Numéro MDL: MFCD00196967
Numéro CAS: 10276-09-2
ADR: 8 III

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR