N-Succinimidyl N-methylcarbamate 97%
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Attention
|
Synonyme(s):
N-Succinimidyl methyllcarbamate, Methylcarbamic acid N-Succinimidyl ester, 2,5-Dioxopyrrolidin-1-yl methylcarbamate
Purity: 97%
CAS No.: 18342-66-0
Molecular Formula: C6H8N2O4
Formula Weight: 172.14
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Sensitivity: Moisture Sensitive
Beilstein Reference: 1531863
MDL No.: MFCD00800292
Formule:
C₆H₈N₂O₄ |
Numéro MDL:
MFCD00800292 Numéro CAS: 18342-66-0 |