4-tert-Butyl-N-(tert-butoxycarbonyl)-L-aspartate 98%
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s):
4-tert-Butyl N-Boc-L-aspartate, N-(tert-Butoxycarbonyl)-L-aspartic Acid 4-tert-Butyl Ester, N-Boc-L-aspartic Acid 4-tert-Butyl Ester, Boc-Asp(OtBu)-OH
Purity: 98%
CAS No.: 1676-90-0
Molecular Formula: C13H23NO6
Formula Weight: 289.32
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Melting Point: 64-67°
Beilstein Reference: 2336144
Optical Rotation: +2°(c=1 in methanol)
MDL No.: MFCD00076912
Formule:
C₁₃H₂₃NO₆ Point de fusion: 64…67 °C |
Numéro MDL:
MFCD00076912 Numéro CAS: 1676-90-0 |