Fmoc-Oic-OH 98%
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Attention
|
Synonyme(s):
(2S,3aS,7aS)-1-Fmoc-octahydroindole-2-carboxylic acid, (2S,3aS,7aS)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)octahydro-1H-indole-2-carboxylic acid
Purity: 98%
CAS No.: 130309-37-4
Molecular Formula: C24H25NO4
Formula Weight: 391.46
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00191201
Formule:
C₂₄H₂₅NO₄ |
Numéro MDL:
MFCD00191201 Numéro CAS: 130309-37-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...