Methyl-2-methylpropionimidate hydrochloride 97%
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Attention
|
Synonyme(s):
Methyl isobutyrimidate hydrochloride
Purity: 97%
CAS No.: 39739-60-1
Molecular Formula: C5H11NO.HCl
Formula Weight: 137.61
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: White
MDL No.: MFCD03844704
Formule:
C₅H₁₂ClNO Poids moléculaire: 137.607 g/mol |
Numéro MDL:
MFCD03844704 Numéro CAS: 39739-60-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...