Glycine benzyl ester-p-toluenesulphonate 99%
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s):
Benzyl glycinate 4-methylbenzenesulphonate, H-Gly-OBzl.Tos-OH, H-Gly-OBzl.TosOH
Purity: 99%
CAS No.: 1738-76-7
Molecular Formula: C9H11NO2.C7H8O3S
Formula Weight: 337.39
Melting Point: 132-134°
Beilstein Reference: 3600116
MDL No.: MFCD00035425
Formule:
C₉H₁₁NO₂ Point de fusion: 132…134 °C Température de stockage: Température ambiante |
Numéro MDL:
MFCD00035425 Numéro CAS: 1738-76-7 EINECS: 217-094-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...