4,6-Dichloropyrimidine-5-carbaldehyde 97%
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Attention
|
Purity: 97%
CAS No.: 5305-40-8
Molecular Formula: C5H2Cl2N2O
Formula Weight: 175.99
Storage Temperature: 2°C to 8°C
Melting Point: 66-71°
MDL No.: MFCD02257701
Formule:
C₅H₂Cl₂N₂O Point de fusion: 66…71 °C |
Numéro MDL:
MFCD02257701 Numéro CAS: 5305-40-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...