Imprimer

N,N'-Dimorpholinomethane 98%

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Danger

H51133.06 H51133.14
H51133.06EA 53.9 EUR
H51133.06 H51133.14
N,N'-Dimorpholinomethane 98%
N,N'-Dimorpholinomethane

Purity: 98% CAS No.: 5625-90-1 Molecular Formula: C9H18N2O2 Formula Weight: 186.25 Boiling Point: 122-124°/12mm MDL No.: MFCD00023369

Formule: C₉H₁₈N₂O₂
Point d'ébullition: 122…124 °C (12 mmHg)
Température de stockage: Température ambiante
Numéro MDL: MFCD00023369
Numéro CAS: 5625-90-1

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR