Methyl-3-aminopropanoate hydrochloride 98%
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s):
beta-Alanine methyl ester hydrochloride, H-β-Ala-OMe·HCl, β-Alanine methyl ester hydrochloride, 3-Aminopropionic acid Methyl Ester Hydrochloride, 3-Aminopropionic Acid Methyl Ester Hydrochloride, Methyl 3-aminopropanoate hydrochloride, Methyl 3-Aminopropionate Hydrochloride, H-β-Ala-OMe.HCl
Purity: 98%
CAS No.: 3196-73-4
Molecular Formula: C4H9NO2.HCl
Formula Weight: 139.58
Storage Temperature: 2°C to 8°C
Melting Point: 102-104°
Beilstein Reference: 3556748
MDL No.: MFCD00039060
Formule:
C₄H₉NO₂·HCl Poids moléculaire: 139.6 g/mol Point de fusion: 102…104 °C |
Numéro MDL:
MFCD00039060 Numéro CAS: 3196-73-4 |