Imprimer
Yeast nitrogen base, without amino acids, without ammonium sulfate
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
H26271.22 H26271.36
H26271.22EA 271 EUR
H26271.22 H26271.36
Yeast nitrogen base, without amino acids, without ammonium sulfate
Milieux pour microbiologie Milieux pour microbiologie déshydratés
Yeast nitrogen base without amino acids and ammonium sulfate is used for classifying yeasts based on carbohydrate and amino acids requirements. Recommended for use in classifying yeasts based on carbon and nitrogen requirements. Contains all essential nutrients required for the growth of yeasts except amino acids, nitrogen and carbohydrate. Addition of nitrogen and carbon sources is required.
MDL No.: MFCD00217952