Imprimer

6-Chloro-7-deazapurine 98%

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Attention

Synonyme(s): 4-Chloro-7H-pyrrolo[2,3-d]pyrimidine

B25713.03 B25713.06
B25713.03EA 123 EUR
B25713.03 B25713.06
6-Chloro-7-deazapurine 98%
6-Chloro-7-deazapurine

Purity: 98% CAS No.: 3680-69-1 Molecular Formula: C6H4ClN3 Formula Weight: 153.52 Storage Temperature: 2°C to 8°C Melting Point: 190-192° MDL No.: MFCD01686865

Formule: C₆H₄ClN₃
Poids moléculaire: 153.57 g/mol
Point de fusion: 190…192 °C
Numéro MDL: MFCD01686865
Numéro CAS: 3680-69-1

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR