Penta-O-acetyl-ɑ-D-glucopyranose 99%
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s):
1,2,3,4,6-Penta-O-acetyl-α-D-glycopyranose, ɑ-D-Glucose Pentaacetate, 1,2,3,4,6-Penta-O-Acetyl-A-D-Glycopyranose, α-D(+)-Glucose pentaacetate, (2R,3R,4S,5R,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate, Pentaacetyl-ɑ-D-glucose, α-D-Glucose pentaacetate
Purity: 99%
CAS No.: 604-68-2
Molecular Formula: C16H22O11
Formula Weight: 390.34
Melting Point: 109-113°
Beilstein Reference: 98852
Optical Rotation: +102° (c=5 in chloroform)
MDL No.: MFCD00064071
Formule:
C₁₆H₂₂O₁₁ Point de fusion: 109…113 °C Température de stockage: Température ambiante |
Numéro MDL:
MFCD00064071 Numéro CAS: 604-68-2 EINECS: 210-073-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...