Imprimer

trans-Indole-3-acrylic acid ≥98%

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Attention

Synonyme(s): trans-3-Indoleacrylic acid

A12143.03 A12143.06
A12143.03EA 61.5 EUR
A12143.03 A12143.06
trans-Indole-3-acrylic acid ≥98%
trans-Indole-3-acrylic acid

Purity: 98+% CAS No.: 29953-71-7 Molecular Formula: C11H9NO2 Formula Weight: 187.20 Storage Temperature: 2°C to 8°C Melting Point: ca 185°dec. Beilstein Reference: 6317 MDL No.: MFCD00005633

Formule: C₁₁H₉NO₂
Poids moléculaire: 187.20 g/mol
Point de fusion: ∼ 185 °C. (déc.)
Numéro MDL: MFCD00005633
Numéro CAS: 29953-71-7
EINECS: 214-872-7

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR