Polyethylene glycol diacrylate (PEGDA), M.W. 3,400
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Danger
|
Synonyme(s):
PEG hydrogel, Polyethylene glycol diacrylate, Poly(ethylene glycol) diacrylate, PEG diacrylate, PEGDA, PEG bisacrylate
CAS No.: 26570-48-9
Molecular Formula: CH2=COC(OCH2CH2)nOCOC=CH2
Storage Temperature: 2°C to 8°C
Physical Form: Granular solid
Appearance: White to off-white
MDL No.: MFCD00081876
Formule:
CH₂=COC(OCH₂CH₂)nOCOC=CH₂ |
Numéro MDL:
MFCD00081876 Numéro CAS: 26570-48-9 EINECS: 607-960-5 |