Imprimer

Zeolite β, hydrogen

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

45875.14 45875.22 45875.36
45875.14EA 58.8 EUR
45875.14 45875.22 45875.36
Zeolite β, hydrogen
Zeolite beta, hydrogen

CAS No.: 1318-02-1 Molecular Formula: SiO2:Al2O3 Physical Form: Powder, S.A. 620 m2/g, 360:1 mole ratio Appearance: White MDL No.: MFCD00132601


Applications: Zeolite beta is used in isomerization of waxes, and Friedel Crafts reactions (alkylation and acylation)

Formule: SiO₂:Al₂O₃
Température de stockage: Température ambiante
Numéro MDL: MFCD00132601
Numéro CAS: 1318-02-1
EINECS: 215-283-8

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR