Imprimer

Zeolite Y, hydrogen

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

45866.14 45866.22 45866.36
45866.14EA 51.4 EUR
45866.14 45866.22 45866.36
Zeolite Y, hydrogen
Zeolite Y, hydrogen

CAS No.: 1318-02-1 Molecular Formula: SiO2:Al2O3 Physical Form: Powder, S.A. 730 m2/g, 5.1:1 mole ratio Appearance: White MDL No.: MFCD00132601


Applications: Zeolite Y is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other products

Formule: SiO₂:Al₂O₃
Température de stockage: Température ambiante
Numéro MDL: MFCD00132601
Numéro CAS: 1318-02-1
EINECS: 215-283-8

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR