Soldering flux, Liquid
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Danger
|
All purpose flux for general soldering.
Densité:
1.26 Température de stockage: Température ambiante |
Numéro MDL:
MFCD01868519 UN: 1840 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...