Imprimer

Silver behenate

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Synonyme(s): Docosanoic Acid Silver Salt, Behenic Acid Silver Salt, Silver Docosanoate

45494.06 45494.14
45494.06EA 132 EUR
45494.06 45494.14
Silver behenate
Silver behenate

CAS No.: 2489-05-6 Molecular Formula: C22H43O2Ag Formula Weight: 447.44 Physical Form: Powder Sensitivity: Light Sensitive Appearance: White Melting Point: dec. MDL No.: MFCD00059001

Formule: C₂₂H₄₄O₂
Point de fusion: Déc.
Température de stockage: Température ambiante
Numéro MDL: MFCD00059001
Numéro CAS: 2489-05-6
EINECS: 219-641-4

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR