Imprimer
Multi-element plasma standard solution, non metals, Specpure®
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
44267.AE 44267.AP
44267.AEEA 188 EUR
44267.AE 44267.AP
Multi-element plasma standard solution, non metals, Specpure®
Étalons Étalons cationiques Standards multi-éléments
As, B, P, S, Se, Si, Te at concentration 100 µg/ml in 5% HCL with a trace of HF.
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00144997