Imprimer
Grinding pieces, zirconium oxide, yttria stabilised
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
43286.A1 43285.A1 43286.30 43285.30
43286.A1EA 598 EUR
43286.A1 43285.A1 43286.30 43285.30
Grinding pieces, zirconium oxide, yttria stabilised
Perles Billes en céramique
The density of the zirconium oxide beads is 6.02 g/cm³
  • MDL No.: MFCD00145546
  • Yttrium oxide (mole) 3%
  • Hardness 12
  • Total impuroty level < 0.1
Storage at ambient temperatures.