Imprimer
Salcomine [=N,N'-bis(salicylidene)ethylenediiminocobalt(II)] ≥96%
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Attention
Synonyme(s): N,N'-Disalicylalethylenediamine Cobalt(II), Salcomine, N,N'-Bis(salicylidene)ethylenediiminocobalt(II), N,N'-Ethylenebis(salicylideneiminato)cobalt(II)
43174.22 43174.03 43174.14 43174.06
43174.22EA 646 EUR
43174.22 43174.03 43174.14 43174.06
Salcomine [=N,N'-bis(salicylidene)ethylenediiminocobalt(II)] ≥96%
Salcomine [=N,N'-bis(salicylidene)ethylenediiminocobalt(II)]
Purity: 96% CAS No.: 14167-18-1 Molecular Formula: C16H14CoN2O2 Formula Weight: 325.23 Physical Form: Powder Appearance: Black Odour: Odourless MDL No.: MFCD00000009
Formule: C₁₆H₁₆CoN₂O₂
Température de stockage: Température ambiante
Numéro MDL: MFCD00000009
Numéro CAS: 14167-18-1
EINECS: 238-012-5