Imprimer
Silica gel, TLC high purity grade, with gypsum binder and fluorescent indicator
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
42764.30 42764.A1
42764.30EA 132 EUR
42764.30 42764.A1
Silica gel, TLC high purity grade, with gypsum binder and fluorescent indicator
Sorbants pour chromatographie Sorbants TLC
This TLC high purity grade silica gel with gypsum binder and fluorescent indicator is used for thin layer chromatography plate preparation.
  • Insoluble in water
  • pH 6,5 to 7,5