Imprimer
Gel de silice 60 0.060-0.2mm (70-230 mesh), avec indicateur fluorescent
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Attention
42757.18 42757.30 42757.A1 42757.A7
42757.18EA 33.1 EUR
42757.18 42757.30 42757.A1 42757.A7
Gel de silice 60 0.060-0.2mm (70-230 mesh), avec indicateur fluorescent
Gel de silice
Silica gel 60 is used as carriers, processing aids, anti-caking and free-flow agents in animal feed. Defoamer applications such as paint, food, paper, textile and other industrial applications. Synthetic silicon dioxides are used as a rheology control agent in plastics. It is also used in dry column chromatography without applied pressure (gravity pulled).
CAS No.: 7631-86-9 Physical Form: -70+230 Mesh Powder, S.A. 500-600m2/g Odour: Odourless MDL No.: MFCD00011232
Applications: Dry column chromatography without applied pressure (gravity pulled)
Formule: SiO₂
Poids moléculaire: 60.08 g/mol
Point d'ébullition: 2230
Point de fusion: 1710
Température de stockage: Température ambiante
Numéro MDL: MFCD00011232
Numéro CAS: 63231-67-4
EINECS: 231-545-4