Imprimer
Silica gel, TLC high purity grade, without binder
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
42756.30 42756.A1 42756.A7
42756.30EA 125 EUR
42756.30 42756.A1 42756.A7
Silica gel, TLC high purity grade, without binder
Sorbants pour chromatographie Sorbants TLC
This TLC high purity grade silica gel without binder is used for thin layer chromatography plate preparation.
  • Incompatible with oxidizing agents
  • Soluble in alkalies
  • Insoluble in water
  • pH 6 to 7