Imprimer

4,4'-Diaminobiphenyl-2,2'-disulphonic acid hydrate, (∼30% H₂O)

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Attention

Synonyme(s): 2,2'-Benzidinedisulphonic Acid, 4,4'-Diamino-[1,1'-biphenyl]-2,2'-disulphonic acid(contains 30% Water at maximum)

41472.14 41472.22
41472.14EA 76.2 EUR
41472.14 41472.22
4,4'-Diaminobiphenyl-2,2'-disulphonic acid hydrate, (∼30% H₂O)
4,4'-Diaminobiphenyl-2,2'-disulphonic acid hydrate

CAS No.: 117-61-3 Molecular Formula: C12H12N2O6S2.xH2O Formula Weight: 344.36(anhy) Physical Form: Powder Appearance: Dark purple MDL No.: MFCD00041885

Formule: C₁₂H₁₂N₂O₆S₂·xH₂O
Poids moléculaire: 344.36 g/mol (anhy)
Température de stockage: Température ambiante
Numéro MDL: MFCD00041885
Numéro CAS: 117-61-3
EINECS: 204-200-0

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR