Imprimer
Polyéthylèneimine, M.W. M.W. 70,000 30% w/v aqueous solution, branched
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Attention
Synonyme(s): Polyaziridine, Corcat, Polyethylenimine, Polymin P, Polyaziridine Mw 25000, Aziridine, PEI, Polyethylenimine (branched), Ethyleneimine Polymer, Poly(ethyleneimine)
40529.14 40529.22 40529.36
40529.14EA 88.8 EUR
40529.14 40529.22 40529.36
Polyéthylèneimine, M.W. M.W. 70,000 30% w/v aqueous solution, branched
Polyethyleneimine
Acts as a protein precipitant used to purify proteins. It is used as chelating agent and as a scavenger for aldehydes and oxides. Also used in detergents, paper industry, dyes, printing inks and in water treatment.
CAS No.: 9002-98-6 Molecular Formula: (-NHCH2CH2-)x(-N(CH2CH2NH2)CH2CH2-)y Physical Form: Liquid Appearance: Colourless to pale yellow Odour: Odourless MDL No.: MFCD00803910
Applications: A protein precipitant used to purify proteins
Formule: (CH₂CH₂NH)n
Température de stockage: Température ambiante
Numéro MDL: MFCD00084427
Numéro CAS: 9002-98-6