Imprimer

Aluminum, silver coated (19 - 21% Ag) (p/p) ≥99.9% (base métaux), en poudre -200 mesh

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

40340.14 40340.22
40340.14EA 88.3 EUR
40340.14 40340.22
Aluminum, silver coated (19 - 21% Ag) (p/p) ≥99.9% (base métaux), en poudre -200 mesh
Aluminum, silver coated

Purity: 99.9% Physical Form: Density 1.1-1.7g/cm3 Odour: Odourless MDL No.: MFCD05865335


Applications: Conductive adhesives

Température de stockage: Température ambiante
Numéro MDL: MFCD05865335
UN: 3077
ADR: 9 III

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR