Imprimer
Ceramic tubes, one end closed
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
30062.DR 30062.DW 30062.EH 30062.EM 30062.EO 30073.DR 30080.DM 30080.DW 30080.EH
30062.DREA 113 EUR
30062.DR 30062.DW 30062.EH 30062.EM 30062.EO 30073.DR 30080.DM 30080.DW 30080.EH
Ceramic tubes, one end closed
Tuyaux Transfer Tubing
Pure Al₂O₃ Aluminium oxide AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.