Cerium(IV) oxide, hydrated
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Danger
|
Synonyme(s):
Cerium(IV) hydroxide
CAS No.: 12014-56-1
Molecular Formula: CeO2.xH2O
Formula Weight: 172.12(anhy)
Physical Form: Lump/Powder
Appearance: Yellow to pale brown
Odour: Odourless
UN No.: UN3262
MDL No.: MFCD00010927
Formule:
CeO₂.xH₂O Température de stockage: Température ambiante |
Numéro MDL:
MFCD00010927 Numéro CAS: 12014-56-1 EINECS: 234-599-7 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...