Imprimer
Aluminum silicate
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): Kaolinite, Silicic acid (H6Si2O7), Aluminum salt (1:2) dihydrate, Aluminum hydroxide silicate (Al2(OH)4(Si2O5)), Aluminum hydroxide silicate (2:4:1), Dialuminum trioxido-Trioxidosilyloxy-silane dihydrate
14231.36 14231.A3
14231.36EA 62.3 EUR
14231.36 14231.A3
Aluminum silicate
Aluminum silicate
Aluminum silicate is used to prepare aluminum-silicate polymer composite (PASiC), which is an inorganic coagulant used in water treatment. It is used as a catalyst for the pyrolysis of rice husk to crude bio -oil, which upgraded to supercritical ethanol. Further, it is used as filler in paper, rubber and in paints.
CAS No.: 12428-46-5 Molecular Formula: Al2Si2O7.2H2O Formula Weight: 258.16 (222.13anhy) Physical Form: Powder Appearance: White to pale grey Odour: Odourless MDL No.: MFCD00062311
Formule: Al₂Si₂O₇·2H₂O
Poids moléculaire: 258.16 g/mol (222.13 anhy)
Température de stockage: Température ambiante
Numéro MDL: MFCD00062311
Numéro CAS: 12428-46-5